ENA161173404 Enamine

N-[(2-bromophenyl)methyl]cyclopropanamine

  • MDL number MFCD09808799
  • Cas Number : 90868-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32123
Availability details for
Quantity
  • Availability
  • Add to Cart