ENA161173409 Enamine

3-(4-nitrophenyl)-1H-1,2,4-triazole

  • MDL number MFCD06090835
  • Cas Number : 6219-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35597
Availability details for
Quantity
  • Availability
  • Add to Cart