ENA161173416 Enamine

2-(4-aminophenyl)-N-ethylacetamide

  • MDL number MFCD09043009
  • Cas Number : 107966-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117...119
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33573

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33573
Availability details for
Quantity
  • Availability
  • Add to Cart