ENA161173422 Enamine

4-(1H-pyrazol-1-yl)-2-(trifluoromethyl)aniline

  • MDL number MFCD09046281
  • Cas Number : 926243-93-8
  • Molecular Weight : 227.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-35616

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35616
Availability details for
Quantity
  • Availability
  • Add to Cart