ENA161173427 Enamine

1-(2,6-dichlorophenyl)pyrrolidin-2-imine hydrochloride

  • MDL number MFCD03091071
  • Cas Number : 1092707-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35622
Availability details for
Quantity
  • Availability
  • Add to Cart