ENA161173438 Enamine

(1-amino-3-phenylpropan-2-yl)diethylamine

  • MDL number MFCD09742781
  • Cas Number : 954565-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35636
Availability details for
Quantity
  • Availability
  • Add to Cart