ENA161173441 Enamine

4-(3-aminopropyl)-N,N-dimethylaniline

  • MDL number MFCD09886638
  • Cas Number : 112103-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35639
Availability details for
Quantity
  • Availability
  • Add to Cart