ENA161173442 Enamine

ethyl 2-(3-chloropropanamido)-5-methyl-4-phenylthiophene-3-carboxylate

  • MDL number MFCD03964526
  • Cas Number : 497091-41-5
  • Molecular Weight : 351.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-17106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-17106
Availability details for
Quantity
  • Availability
  • Add to Cart