ENA161173445 Enamine

2-{[5-(trifluoromethyl)pyridin-2-yl]amino}ethanol

  • MDL number MFCD07359872
  • Cas Number : 874630-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35644

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35644
Availability details for
Quantity
  • Availability
  • Add to Cart