ENA161173447 Enamine

2-(methoxymethyl)benzoic acid

  • MDL number MFCD09863839
  • Cas Number : 88550-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-92
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35646

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35646
Availability details for
Quantity
  • Availability
  • Add to Cart