ENA161173449 Enamine

5-(propan-2-yl)-1-[5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD10686729
  • Cas Number : 1135324-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35648
Availability details for
Quantity
  • Availability
  • Add to Cart