ENA161173459 Enamine

[methyl(pyrimidin-2-yl)amino]acetic acid

  • MDL number MFCD09733226
  • Cas Number : 953904-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33619

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33619
Availability details for
Quantity
  • Availability
  • Add to Cart