ENA161173463 Enamine

2-(4-aminophenyl)-N,N-diethylacetamide

  • MDL number MFCD09047822
  • Cas Number : 887479-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35681

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35681
Availability details for
Quantity
  • Availability
  • Add to Cart