ENA161173464 Enamine

3-carbamoyl-4-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD09049243
  • Cas Number : 76503-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35682
Availability details for
Quantity
  • Availability
  • Add to Cart