ENA161173471 Enamine

4-(aminomethyl)-N-isopropyl-N-methylbenzamide

  • MDL number MFCD08444574
  • Cas Number : 923233-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35690

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35690
Availability details for
Quantity
  • Availability
  • Add to Cart