ENA161173476 Enamine

5-acetyl-3-methylthieno[2,3-b]pyridine-2-carboxylic acid

  • MDL number MFCD10664483
  • Cas Number : 1050885-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35696

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35696
Availability details for
Quantity
  • Availability
  • Add to Cart