ENA161173485 Enamine

2-[4-(2-methyl-1,3-thiazol-4-yl)phenyl]acetic acid

  • MDL number MFCD10001835
  • Cas Number : 77015-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35709
Availability details for
Quantity
  • Availability
  • Add to Cart