ENA161173487 Enamine

3-(2,6-dimethylphenoxy)propanoic acid

  • MDL number MFCD09044373
  • Cas Number : 22383-95-5
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35711

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35711
Availability details for
Quantity
  • Availability
  • Add to Cart