ENA161173514 Enamine

2-fluoro-4-phenylaniline hydrochloride

  • MDL number MFCD10666929
  • Cas Number : 1171915-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-33636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33636
Availability details for
Quantity
  • Availability
  • Add to Cart