ENA161173518 Enamine

1-[2-(2,2,2-trifluoroethoxy)pyridin-3-yl]methanamine

  • MDL number MFCD06212927
  • Cas Number : 771581-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35761
Availability details for
Quantity
  • Availability
  • Add to Cart