ENA161173521 Enamine

6-(1-aminoethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD08444636
  • Cas Number : 159459-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35764
Availability details for
Quantity
  • Availability
  • Add to Cart