ENA161173525 Enamine

3-amino-4-chloro-N,N-dimethylbenzamide

  • MDL number MFCD09732653
  • Cas Number : 51919-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35772
Availability details for
Quantity
  • Availability
  • Add to Cart