ENA161173528 Enamine

N-[(oxolan-2-yl)methyl]thiolan-3-amine

  • MDL number MFCD10686760
  • Cas Number : 1021130-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35775
Availability details for
Quantity
  • Availability
  • Add to Cart