ENA161173531 Enamine

4-(aminocarbonyl)cyclohexanecarboxylic acid

  • MDL number MFCD09814965
  • Cas Number : 55794-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205...207
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35778

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35778
Availability details for
Quantity
  • Availability
  • Add to Cart