ENA161173555 Enamine

3-{1,3,4-trimethyl-6-oxo-1H,6H,7H-pyrazolo[3,4-b]pyridin-5-yl}propanoic acid

  • MDL number MFCD10666940
  • Cas Number : 1049605-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33661
Availability details for
Quantity
  • Availability
  • Add to Cart