ENA161173567 Enamine

6-chloro-3,4-dihydro-2H-1,4-benzothiazine hydrochloride

  • MDL number MFCD10666943
  • Cas Number : 1171671-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-33669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33669
Availability details for
Quantity
  • Availability
  • Add to Cart