ENA161173573 Enamine

(1H-1,3-benzodiazol-2-yl)(phenyl)methanol

  • MDL number MFCD00159953
  • Cas Number : 50-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-211
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-33676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33676
Availability details for
Quantity
  • Availability
  • Add to Cart