ENA161208451 Enamine

2-chloro-5-methanesulfonylaniline hydrochloride

  • MDL number MFCD08447122
  • Cas Number : 1170556-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-74
Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-04060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04060
Availability details for
Quantity
  • Availability
  • Add to Cart