ENA161208452 Enamine

N-(2,5-dimethylphenyl)-2-(methylamino)acetamide hydrochloride

  • MDL number MFCD08447229
  • Cas Number : 1171227-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202-203
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-04249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04249
Availability details for
Quantity
  • Availability
  • Add to Cart