ENA161208469 Enamine

5-chloro-2-(4-methylphenoxy)aniline hydrochloride

  • MDL number MFCD08447205
  • Cas Number : 1173051-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06473

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06473
Availability details for
Quantity
  • Availability
  • Add to Cart