ENA161208485 Enamine

4-{2,5-dimethyl-1-[3-(trifluoromethyl)phenyl]-1H-pyrrol-3-yl}-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD08447175
  • Cas Number : 1172928-96-9
  • Molecular Weight : 373.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07698

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-07698
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart