ENA161208486 Enamine

4-[1-(1,3-benzodioxol-5-yl)-2,5-dimethyl-1H-pyrrol-3-yl]-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD08447191
  • Cas Number : 1172446-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240...242
Purity 95.00
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07699

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07699
Availability details for
Quantity
  • Availability
  • Add to Cart