ENA161208489 Enamine

N-(2,6-dimethylphenyl)-2-(methylamino)acetamide hydrochloride

  • MDL number MFCD08447232
  • Cas Number : 35891-84-0
  • Molecular Weight : 228.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07822
Availability details for
Quantity
  • Availability
  • Add to Cart