ENA161208493 Enamine

N-(4-methoxyphenyl)-2-(methylamino)acetamide hydrochloride

  • MDL number MFCD08447241
  • Cas Number : 1171427-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208-209
Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07828

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07828
Availability details for
Quantity
  • Availability
  • Add to Cart