ENA161208527 Enamine

4-{1-[(furan-2-yl)methyl]-2,5-dimethyl-1H-pyrrol-3-yl}-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD08447195
  • Cas Number : 1171580-10-1
  • Molecular Weight : 309.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-10928

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10928
Availability details for
Quantity
  • Availability
  • Add to Cart