ENA161208569 Enamine

[(6-bromo-1,3-dioxaindan-5-yl)methyl]hydrazine hydrochloride

  • MDL number MFCD09834101
  • Cas Number : 1171184-22-7
  • Molecular Weight : 281.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29450

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29450
Availability details for
Quantity
  • Availability
  • Add to Cart