ENA161208574 Enamine

(2,2-dimethylpropyl)hydrazine dihydrochloride

  • MDL number MFCD09834107
  • Cas Number : 1177323-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29517
Availability details for
Quantity
  • Availability
  • Add to Cart