ENA161208576 Enamine

1-[1-(3-benzyl-1,2,4-oxadiazol-5-yl)ethyl]piperazine hydrochloride

  • MDL number MFCD10686677
  • Cas Number : 1156900-49-0
  • Molecular Weight : 308.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-31670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31670
Availability details for
Quantity
  • Availability
  • Add to Cart