ENA161208579 Enamine

methyl 1-(4-aminophenyl)-1H-1,2,3-triazole-4-carboxylate

  • MDL number MFCD10686730
  • Cas Number : 1049605-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35652
Availability details for
Quantity
  • Availability
  • Add to Cart