ENA161208583 Enamine

4-[1-(2,3-dihydro-1,4-benzodioxin-6-yl)-2,5-dimethyl-1H-pyrrol-3-yl]-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD07286006
  • Cas Number : 1049777-13-0
  • Molecular Weight : 363.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07283

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-07283
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart