ENA161208598 Enamine

3-(butylamino)-1lambda6-thiolane-1,1-dione hydrochloride

  • MDL number MFCD00456576
  • Cas Number : 76924-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07603
Availability details for
Quantity
  • Availability
  • Add to Cart