ENA161208605 Enamine

4-(2,5-dimethyl-1-phenyl-1H-pyrrol-3-yl)-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD07308583
  • Cas Number : 1052550-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07694

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07694
Availability details for
Quantity
  • Availability
  • Add to Cart