ENA161208606 Enamine

4-[2,5-dimethyl-1-(4-methylphenyl)-1H-pyrrol-3-yl]-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD07286291
  • Cas Number : 1049726-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-07695

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07695
Availability details for
Quantity
  • Availability
  • Add to Cart