ENA161208618 Enamine

[(2-chloro-6-fluorophenyl)methyl](methyl)amine hydrochloride

  • MDL number MFCD07106786
  • Cas Number : 1158252-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-136
Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07735

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07735
Availability details for
Quantity
  • Availability
  • Add to Cart