ENA161208620 Enamine

2-(methylamino)-N-(2,3,4-trifluorophenyl)acetamide hydrochloride

  • MDL number MFCD07286586
  • Cas Number : 1049764-31-9
  • Molecular Weight : 254.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07796
Availability details for
Quantity
  • Availability
  • Add to Cart