ENA161208621 Enamine

4-(chloromethyl)-N-(2-methylphenyl)-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD07280414
  • Cas Number : 1049751-56-5
  • Molecular Weight : 275.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-02008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02008
Availability details for
Quantity
  • Availability
  • Add to Cart