ENA161208623 Enamine

N-(3-methoxyphenyl)-2-(methylamino)acetamide hydrochloride

  • MDL number MFCD07286583
  • Cas Number : 1049764-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156...158
Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07799

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07799
Availability details for
Quantity
  • Availability
  • Add to Cart