ENA161208624 Enamine

4-(chloromethyl)-2-(4-ethoxyphenyl)-1,3-thiazole hydrochloride

  • MDL number MFCD07286596
  • Cas Number : 1052537-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07816

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07816
Availability details for
Quantity
  • Availability
  • Add to Cart