ENA161208632 Enamine

N1-(2-phenylethyl)benzene-1,2-diamine hydrochloride

  • MDL number MFCD07276079
  • Cas Number : 1189984-54-0
  • Molecular Weight : 248.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-02085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02085
Availability details for
Quantity
  • Availability
  • Add to Cart