ENA161208633 Enamine

N-(3,4-diethoxyphenyl)-2-(methylamino)acetamide hydrochloride

  • MDL number MFCD07286594
  • Cas Number : 1049790-01-3
  • Molecular Weight : 288.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07833
Availability details for
Quantity
  • Availability
  • Add to Cart